Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Lymphotoxin beta/TNFSF3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1684
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1684 500 μg
MAB1684SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1684 Supplier R&D Systems Supplier No. MAB1684
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

Lymphotoxin beta/TNFSF3 Monoclonal specifically detects Lymphotoxin beta/TNFSF3 in Human samples. It is validated for Western Blot.

Specifications

Antigen Lymphotoxin beta/TNFSF3
Applications Western Blot
Classification Monoclonal
Clone 135105
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias LTB, lymphotoxin beta (TNF superfamily, member 3), lymphotoxin-beta, p33, TNFCTNF-C, TNFSF3, TNFSF3LT-beta, Tumor necrosis factor C, Tumor necrosis factor ligand superfamily member 3
Gene Symbols LTB
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Lymphotoxin alpha/beta
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4050
Test Specificity Detects human Lymphotoxin alpha beta heterotrimers in direct ELISAs and Western blots. This antibody detects an epitope in the Lymphotoxin beta subunit. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) Lymphotoxin alpha , rhAPRIL, rhBAFF, rhTNF-alpha , rhTWEAK, rhEDA-A2, rhEDA, rhTRAIL, rhVEGI, rhTRANCE, rhFas Ligand, or rhOX40 Ligand.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.