Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LYPD1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6855
This item is not returnable.
View return policy
Description
LYPD1 Polyclonal specifically detects LYPD1 in Human samples. It is validated for Immunohistochemistry.Specifications
LYPD1 | |
Polyclonal | |
Unconjugated | |
Q8N2G4 | |
LYPD1 | |
E. coli-derived recombinant human LYPD1 Cys54-Ser116 Accession # Q8N2G4 | |
100 μg | |
Primary | |
Detects human LYPD1 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human LYPD3 is observed. | |
Human | |
IgG |
Immunohistochemistry | |
LYOPH | |
Immunohistochemistry 5-15 ug/mL | |
FLJ41033, LY6/PLAUR domain containing 1, ly6/PLAUR domain-containing protein 1, LYPDC1, MGC29643, PHTS, Putative HeLa tumor suppressor | |
Sheep | |
Affinity Purified | |
RUO | |
116372 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction