Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LYPD8 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9087T100UG
This item is not returnable.
View return policy
Description
LYPD8 Monoclonal specifically detects LYPD8 in Human samples. It is validated for Flow Cytometry.Specifications
LYPD8 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human LYPD8 in direct ELISAs. Stains human LYPD8 transfectants but not irrelevant transfectants in flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Flow Cytometry | |
961703 | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q6UX82 | |
Human embryonic kidney cell line HEK293-derived transfected with human LYPD8, Met1-Asn215, Accession # Q6UX82 | |
100 μg | |
646627 | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction