Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Lysosomal Pro-X Carboxypeptidase/PRCP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7164
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7164 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7164 Supplier R&D Systems Supplier No. MAB7164
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Lysosomal Pro-X Carboxypeptidase/PRCP Monoclonal specifically detects Lysosomal Pro-X Carboxypeptidase/PRCP in Human samples. It is validated for Western Blot.

Specifications

Antigen Lysosomal Pro-X Carboxypeptidase/PRCP
Applications Western Blot
Classification Monoclonal
Clone 860821
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P42785
Gene Alias Angiotensinase C, EC 3.4.16.2, HUMPCP, Lysosomal carboxypeptidase C, lysosomal Pro-X carboxypeptidase, PCPMGC2202, Proline carboxypeptidase, Prolylcarboxypeptidase, prolylcarboxypeptidase (angiotensinase C), prolylcarboxypeptidase isoform 1 preproprotein
Gene Symbols PRCP
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Lysosomal Pro-X Carboxypeptidase/PRCP Met1-His496 Accession # P42785
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5547
Test Specificity Detects human Lysosomal Pro-X Carboxypeptidase/PRCP in ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.