Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Lysosomal Pro-X Carboxypeptidase/PRCP Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7164SP
This item is not returnable.
View return policy
Description
Lysosomal Pro-X Carboxypeptidase/PRCP Monoclonal specifically detects Lysosomal Pro-X Carboxypeptidase/PRCP in Human samples. It is validated for Western Blot.Specifications
Lysosomal Pro-X Carboxypeptidase/PRCP | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P42785 | |
PRCP | |
Mouse myeloma cell line NS0-derived recombinant human Lysosomal Pro-X Carboxypeptidase/PRCP Met1-His496 Accession # P42785 | |
25 μg | |
Primary | |
Detects human Lysosomal Pro-X Carboxypeptidase/PRCP in ELISAs. | |
Human | |
Purified |
Western Blot | |
860821 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Angiotensinase C, EC 3.4.16.2, HUMPCP, Lysosomal carboxypeptidase C, lysosomal Pro-X carboxypeptidase, PCPMGC2202, Proline carboxypeptidase, Prolylcarboxypeptidase, prolylcarboxypeptidase (angiotensinase C), prolylcarboxypeptidase isoform 1 preproprotein | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5547 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction