Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human M-CSF R PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB329P100
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB329P100 100 Tests
FAB329P025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB329P100 Supplier R&D Systems Supplier No. FAB329P100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

M-CSFR/CD115 Monoclonal specifically detects M-CSFR/CD115 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen M-CSFR/CD115
Applications Flow Cytometry
Classification Monoclonal
Clone 61708
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P07333.2
Gene Alias CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF1R, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, MCSFR, M-CSFR, Proto-oncogene c-Fms
Gene Symbols CSF1R
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human M-CSF R Ile20-Glu512 (Pro54Ala) Accession # P07333.2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1436
Test Specificity Detects human M-CSF R in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) GM-CSF R alpha or rhGM-CSF R beta .
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.