Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MAdCAM-1 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6056U100UG
This item is not returnable.
View return policy
Description
MAdCAM-1 Monoclonal specifically detects MAdCAM-1 in Human samples. It is validated for Flow Cytometry.Specifications
MAdCAM-1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAY82472 | |
MADCAM1 | |
Mouse myeloma cell line NS0-derived recombinant human MAdCAM-1 Leu21-Gln333 Accession # AAY82472 | |
RUO | |
8174 | |
Human | |
IgG1 |
Flow Cytometry | |
683715 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
hMAdCAM-1, MACAM1, MAdCAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1 | |
Mouse | |
100 μg | |
Primary | |
Detects human MAdCAM-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse MAdCAM-1, recombinant human (rh) ALCAM, rhBCAM, rhCEACAM-1, rhEpCAM, rhICAM-1, -2, -3, -4, -5, rhCD31/PECAM-1, or rhVCAM-1 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction