Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MAdCAM-1 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6056S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6056S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6056S100 Supplier R&D Systems Supplier No. FAB6056S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAdCAM-1 Monoclonal specifically detects MAdCAM-1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen MAdCAM-1
Applications Flow Cytometry
Classification Monoclonal
Clone 683715
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. AAY82472
Gene Alias hMAdCAM-1, MACAM1, MAdCAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1
Gene Symbols MADCAM1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MAdCAM-1 Leu21-Gln333 Accession # AAY82472
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8174
Test Specificity Detects human MAdCAM-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse MAdCAM-1, recombinant human (rh) ALCAM, rhBCAM, rhCEACAM-1, rhEpCAM, rhICAM-1, -2, -3, -4, -5, rhCD31/PECAM-1, or rhVCAM-1 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.