Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MADD Synthetic Peptide

Catalog No. PEP0012
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0012 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0012 Supplier Invitrogen™ Supplier No. PEP0012
Add to Cart
Add to Cart

Synthetic Peptide

MADD Synthetic Peptide, PEP-0012, from Invitrogen.

Tumor necrosis factor receptor 1 (TNFR1) contains a death receptor domain that mediates interactions involved in downstream signaling of TNF. A novel protein, MADD (mitogen activated protein (MAP)-kinase activating death domain protein) has recently been shown to associate with the death domain of TNFR1 through its own C-terminal death domain. This interaction is critical for TNFR1 signal generation. Overexpression of MADD activates the MAP (Mitogen Activated Protein) kinases extracellular signal-related kinase (ERK) and c-Jun N-terminal kinase (JNK) and induces the phosphorylation of cytosolic phospholipase A2. This discovery suggests that MADD links tumor necrosis factor TNF and MAP kinase activation and arachidonic acid release. MADD has a 98% homology with another protein, differentially expressed in neoplasmic vs. normal cells (DENN). DENN is a substrate for JNK3, and is important in the hypoxia and stress induced intracellular signalling pathways. MADD also has a 93% sequence conservation with a GDP/GTP exchange protein, Rab3.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 8567
Species Human
Purification Method purified
Accession Number Q8WXG6
Common Name Human MADD
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol MADD
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.