Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MAGEC1 Partial ORF (NP_005453.2, 848 a.a. - 918 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89951472
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene is a member of the melanoma antigen gene (MAGE) family. The proteins of this family are tumor-specific antigens that can be recognized by autologous cytolytic T lymphocytes. This protein contains a large number of unique short repetitive sequences in front of the MAGE-homologous sequence, and therefore is about 800 aa longer than the other MAGE proteins. [provided by RefSeq]

Sequence: ESPLQSPVISFSSSTSLSPFSEESSSPVDEYTSSSDTLLESDSLTDSESLIESEPLFTYTLDEKVDELARF

Specifications

Accession Number NP_005453.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9947
Molecular Weight (g/mol) 33.44kDa
Name MAGEC1 (Human) Recombinant Protein (Q03)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen ESPLQSPVISFSSSTSLSPFSEESSSPVDEYTSSSDTLLESDSLTDSESLIESEPLFTYTLDEKVDELARF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CT7/MAGE-C1/MGC39366
Common Name MAGEC1
Gene Symbol MAGEC1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.