Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MAGED2 Full-length ORF (NP_055414.2, 1 a.a. - 606 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89951482
Click to view available options
:
2μg

Used for AP, Array, ELISA, WB-Re

This gene is a member of the MAGED gene family. While the MAGEA and MAGEB genes are silent in normal tissues with the exception of testis and placenta, the MAGED genes are expressed ubiquitously. The MAGED genes are clustered on chromosome Xp11. This gene is located in Xp11.2, a hot spot for X-linked mental retardation (XLMR). Multiple alternatively spliced transcript variants have been found for this gene, however, the full length nature of some variants has not been defined. [provided by RefSeq]

Sequence: MSDTSESGAGLTRFQAEASEKDSSSMMQTLLTVTQNVEVPETPKASKALEVSEDVKVSKASGVSKATEVSKTPEAREAPATQASSTTQLTDTQVLAAENKSLAADTKKQNADPQAVTMPATETKKVSHVADTKVNTKAQETEAAPSQAPADEPEPESAAAQSQENQDTRPKVKAKKARKVKHLDGEEDGSSDQSQASGTTGGRRVSKALMASMARRASRGPIAFWARRASRTRLAAWARRALLSLRSPKARRGKARRRAAKLQSSQEPEAPPPRDVALLQGRANDLVKYLLAKDQTKIPIKRSDMLKDIIKEYTDVYPEIIERAGYSLEKVFGIQLKEIDKNDHLYILLSTLEPTDAGILGTTKDSPKLGLLMVLLSIIFMNGNRSSEAVIWEVLRKLGLRPGIHHSLFGDVKKLITDEFVKQKYLDYARVPNSNPPEYEFFWGLRSYYETSKMKVLKFACKVQKKDPKEWAAQYREAMEADLKAAAEAAAEAKARAEIRARMGIGLGSENAAGPCNWDEADIGPWAKARIQAGAEAKAKAQESGSASTGASTSTNNSASASASTSGGFSAGASLTATLTFGLFAGLGGAGASTSGSSGACGFSYK

Specifications

Accession Number NP_055414.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10916
Molecular Weight (g/mol) 91.4kDa
Name MAGED2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 ug
Immunogen MSDTSESGAGLTRFQAEASEKDSSSMMQTLLTVTQNVEVPETPKASKALEVSEDVKVSKASGVSKATEVSKTPEAREAPATQASSTTQLTDTQVLAAENKSLAADTKKQNADPQAVTMPATETKKVSHVADTKVNTKAQETEAAPSQAPADEPEPESAAAQSQENQDTRPKVKAKKARKVKHLDGEEDGSSDQSQASGTTGGRRVSKALMASMARRASRGPIAFWARRASRTRLAAWARRALLSLRSPKARRGKARRRAAKLQSSQEPEAPPPRDVALLQGRANDLVKYLLAKDQTKIPIKRSDMLKDIIKEYTDVYPEIIERAGYSLEKVFGIQLKEIDKNDHLYILLSTLEPTDAGILGTTKDSPKLGLLMVLLSIIFMNGNRSSEAVIWEVLRKLGLRPGIHHSLFGDVKKLITDEFVKQKYLDYARVPNSNPPEYEFFWGLRSYYETSKMKVLKFACKVQKKDPKEWAAQYREAMEADLKAAAEAAAEAKARAEIRARMGIGLGSENAAGPCNWDEADIGPWAKARIQAGAEAKAKAQESGSASTGASTSTNNSASASASTSGGFSAGASLTATLTFGLFAGLGGAGASTSGSSGACGFSYK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias 11B6/BCG1/HCA10/JCL-1/MAGE-D2/MAGED/MGC8386
Common Name MAGED2
Gene Symbol MAGED2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.