Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MAGP-2/MFAP5 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems BAF4914
This item is not returnable.
View return policy
Description
MAGP-2/MFAP5 Polyclonal specifically detects MAGP-2/MFAP5 in Human samples. It is validated for Western Blot.Specifications
MAGP-2/MFAP5 | |
Polyclonal | |
Biotin | |
MAGP2, MAGP-2, MAGP2MFAP-5, MFAP5, Microfibril-associated glycoprotein 2, microfibrillar associated protein 5, microfibrillar-associated protein 5, MP25, MP25microfibril-associated glycoprotein-2 | |
Sheep | |
Affinity Purified | |
RUO | |
8076 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
MFAP5 | |
Mouse myeloma cell line NS0-derived recombinant human MAGP-2/MFAP5 | |
50 μg | |
Primary | |
Detects human MAGP-2/MFAP5 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human MAGP-1 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction