Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MAP3K8/Tpl2/COT Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4586SP
This item is not returnable.
View return policy
Description
MAP3K8/Tpl2/COT Monoclonal specifically detects MAP3K8/Tpl2/COT in Human samples. It is validated for Western Blot.Specifications
MAP3K8/Tpl2/COT | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P41279 | |
MAP3K8 | |
E. coli-derived recombinant human MAP3K8/Tpl2/COT Gln173-Gln395 Accession # P41279 | |
25 μg | |
Primary | |
Detects human MAP3K8/Tpl2/COT in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot | |
634130 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cancer Osaka thyroid oncogene, c-COT, COT, cot (cancer Osaka thyroid) oncogene, COTMEKK8, EC 2.7.11.25, EST, ESTF, ESTFLJ10486, Ewing sarcoma transformant, MAP3K8, mitogen-activated protein kinase kinase kinase 8, Proto-oncogene c-Cot, proto-oncogene serine/threoine protein kinase, Serine/threonine-protein kinase cot, Tpl-2, TPL2, Tumor progression locus 2, tumor progression locus-2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1326 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction