Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Marapsin/Pancreasin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1988
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1988 100 μg
AF1988SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1988 Supplier R&D Systems Supplier No. AF1988
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Marapsin/Pancreasin Polyclonal specifically detects Marapsin/Pancreasin in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Marapsin/Pancreasin
Applications Western Blot, Immunoprecipitation, Neutralization, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 2 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Gene Accession No. Q9BQR3
Gene Alias CAP-2, CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPN, MPNMarapsin, Pancreasin, protease, serine 27, PRSS27, serine protease 27
Gene Symbols PRSS27
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Marapsin/Pancreasin Ala23-Lys290 Accession # Q9BQR3
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83886
Test Specificity Detects human Marapsin/Pancreasin in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant mouse Marapsin is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.