Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MASTL Antibody, R&D Systems™

Goat Antibody
$200.50 - $526.50
Specifications
Antigen | MASTL |
---|---|
Dilution | Immunocytochemistry 5-15 μg;/mL |
Applications | Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AF10022SP
![]() |
R&D Systems
AF10022SP |
25 μg |
Each for $200.50
|
|
|||||
AF10022100
![]() |
R&D Systems
AF10022100 |
100 μg |
Each for $526.50
|
|
|||||
Description
MASTL specifically detects MASTL in Human samples. It is validated for Immunocytochemistry.Specifications
MASTL | |
Immunocytochemistry | |
Unconjugated | |
Goat | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
FLJ14813EC 2.7.11.1, GREATWALL, GW, GWL, hGWL, MAST-L, microtubule associated serine/threonine kinase-like, Microtubule-associated serine/threonine-protein kinase-like, RP11-85G18.2, THC2, THC2greatwall protein kinase | |
E. coli-derived recombinant human MASTL., Lys39-Lys158, Accession No. Q96GX5 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human MASTL in direct ELISA. |
Immunocytochemistry 5-15 μg;/mL | |
Polyclonal | |
Lyophilized | |
Human | |
Q96GX5 | |
84930.0 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title