Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Matriptase/ST14 Catalytic Domain Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC3946R100UG
This item is not returnable.
View return policy
Description
Matriptase/ST14 Monoclonal specifically detects Matriptase/ST14 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Matriptase/ST14 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9Y5Y6 | |
ST14 | |
E. coli-derived recombinant human Matriptase/ST14 Catalytic Domain Val615-Val855 Accession # Q9Y5Y6 | |
RUO | |
6768 | |
Human | |
IgG1 |
Flow Cytometry | |
416802 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
EC 3.4.21, Epithin, HAI, Matriptase, Membrane-type serine protease 1, MTSP1, MT-SP1EC 3.4.21.109, prostamin, PRSS14, Serine protease 14, Serine protease TADG-15, SNC19, SNC19MTSP1, ST14, suppression of tumorigenicity 14 (colon carcinoma), suppression of tumorigenicity 14 (colon carcinoma, matriptase, epithin), suppressor of tumorigenicity 14 protein, TADG15, TADG-15, TMPRSS14, tumor associated differentially expressed gene 15 protein, Tumor-associated differentially-expressed gene 15 protein | |
Mouse | |
100 μg | |
Primary | |
Detects human Matriptase/ST14 Catalytic Domain in direct ELISAs and Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction