Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human mCherry Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB11041G100UG
This item is not returnable.
View return policy
Description
mCherry Monoclonal antibody specifically detects mCherry in Human samples. It is validated for Flow CytometrySpecifications
mCherry | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
2783C | |
Flow Cytometry | |
DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein | |
Human mCherry synthetic peptide | |
100 μg | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction