Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MCHR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB79382SP
This item is not returnable.
View return policy
Description
MCHR1 Monoclonal antibody specifically detects MCHR1 in Human samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
MCHR1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
G protein-coupled receptor 24, GPR24, GPR24SLC-1, G-protein coupled receptor 24, MCH receptor 1, MCH1R, MCH-1R, MCH1RMCHR-1, MCH-R1, melanin-concentrating hormone receptor 1, MGC32129, SLC1, SLC1MCHR, Somatostatin receptor-like protein | |
E. coli-derived human MCHR1, Met1-Pro113, Accession # Q99705 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG1 |
Immunohistochemistry, Immunocytochemistry | |
1032150 | |
Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q99705 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2847 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction