Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Mcl-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF8281SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8281SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF8281SP Supplier R&D Systems Supplier No. AF8281SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Mcl-1 Polyclonal specifically detects Mcl-1 in Human samples. It is validated for Western Blot, Knockout Validated.

Specifications

Antigen Mcl-1
Applications Western Blot, Immunoassay
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q07820
Gene Alias bcl2-L-3, BCL2L3, BCL2L3MGC104264, Bcl-2-like protein 3, Bcl-2-related protein EAT/mcl1, EAT, induced myeloid leukemia cell differentiation protein Mcl-1, Mcl1, Mcl-1, mcl1/EAT, MCL1-ES, MCL1L, MCL1S, MGC1839, myeloid cell leukemia ES, myeloid cell leukemia sequence 1 (BCL2-related), TM
Gene Symbols MCL1
Host Species Sheep
Immunogen E. coli-derived recombinant human Mcl-1 Val147-Gly219 Accession # Q07820
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4170
Test Specificity Detects human Mcl-1 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.