Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MCT1/SLC16A1 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8275T100UG
This item is not returnable.
View return policy
Description
MCT1/SLC16A1 Monoclonal specifically detects MCT1/SLC16A1 in Human samples. It is validated for Flow Cytometry.Specifications
MCT1/SLC16A1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P53985 | |
SLC16A1 | |
NS0 mouse myeloma cell line transfected with human MCT1/SLC16A1 Met1-Val500 Accession # P53985 | |
RUO | |
6566 | |
Human | |
IgG2a |
Flow Cytometry | |
882616 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
FLJ36745, HHF7, MCT, MCT 1, MCT1MGC44475, member 1, monocarboxylate transporter 1, SLC16A1, Solute carrier family 16 member 1, solute carrier family 16, member 1 (monocarboxylic acid transporter 1) | |
Mouse | |
100 μg | |
Primary | |
Stains human MCT1/SLC16A1 transfected cells but not irrelevant transfectants in flow cytometry. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction