Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MD-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB925
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB925 500 μg
MAB925SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB925 Supplier R&D Systems Supplier No. MAB925
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MD-1/LY86 Monoclonal specifically detects MD-1/LY86 in Human samples. It is validated for Western Blot.

Specifications

Antigen MD-1/LY86
Applications Western Blot
Classification Monoclonal
Clone 153014
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O95711
Gene Alias Ly-86, LY86, lymphocyte antigen 86, MD1, MMD-1, Protein MD-1, RP105-associated
Gene Symbols LY86
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MD-1 Gly21-Ser162 Accession # O95711
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9450
Test Specificity Detects human MD-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse MD-1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.