Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MD2 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems BAF1787
This item is not returnable.
View return policy
Description
MD-2 Polyclonal specifically detects MD-2 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
MD-2 | |
Polyclonal | |
Biotin | |
NP_001123946 | |
LY96 | |
E. coli-derived recombinant human MD-2 Glu17-Asn160 Accession # NP_001123946 | |
50 μg | |
Primary | |
Detects human MD-2 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human MD-1 is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
ESOP1, ESOP-1, Ly-96, LY96, lymphocyte antigen 96, MD2, MD-2, myeloid differentiation protein-2, Protein MD-2 | |
Sheep | |
Affinity Purified | |
RUO | |
23643 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction