Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MDL-1/CLEC5A Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2384T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2384T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2384T100 Supplier R&D Systems Supplier No. FAB2384T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MDL-1/CLEC5A Monoclonal specifically detects MDL-1/CLEC5A in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen MDL-1/CLEC5A
Applications Flow Cytometry
Classification Monoclonal
Clone 283834
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9NY25
Gene Alias CLEC5A, CLECSF5, CLECSF5carbohydrate-recognition domain) lectin, superfamilymember 5, C-type lectin domain family 5, member A, C-type lectin superfamily member 5, MDL1, MDL-1MGC138304, Myeloid DAP12-associating lectin 1, myeloid DAP12-associating lectin-1
Gene Symbols CLEC5A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MDL-1/CLEC5A Tyr26-Lys188 Accession # Q9NY25
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23601
Test Specificity Detects human MDL-1/CLEC5A in Western blots. Does not cross-react with recombinant mouse MDL-1.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.