Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MED17 Full-length ORF (AAH21101.1, 1 a.a. - 651 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89940582
Click to view available options
Quantity:
2 μg

Used for AP, Array, ELISA, WB-Re

The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. [provided by RefSeq]

Sequence: MSGVRAVRISIESACEKQVHEVGLDGTETYLPPLSMSQNLARLAQRIDFSQGSGSEEEEAAGTEGDAQDWPGAGSSADQDDEEGVVKFQPSLWPWDSVRNNLRSALTEMCVLYDVLSIVRDKKFMTLDPVSQDALPPKQNPQTLQLISKKKSLAGAAQILLKGAERLTKSVTENQENKLQRDFNSELLRLRQHWKLRKVGDKILGDLSYRSAGSLFPHHGTFEVIKNTDLDLDKKIPEDYCPLDVQIPSDLEGSAYIKVSIQKQAPDIGDLGTVNLFKRPLPKSKPGSPHWQTKLEAAQNVLLCKEIFAQLSREAVQIKSQVPHIVVKNQIISQPFPSLQLSISLCHSSNDKKSQKFATEKQCPEDHLYVLEHNLHLLIREFHKQTLSSIMMPHPASAPFGHKRMRLSGPQAFDKNEINSLQSSEGLLEKIIKQAKHIFLRSRAAATIDSLASRIEDPQIQAHWSNINDVYESSVKVLITSQGYEQICKSIQLQLNIGVEQIRVVHRDGRVITLSYQEQELQDFLLSQMSQHQVHAVQQLAKVMGWQVLSFSNHVGLGPIESIGNASAITVASPSGDYAISVRNGPESGSKIMVQFPRNQCKDLPKSDVLQDNKWSHLRGPFKEVQWNKMEGRNFVYKMELLMSALSPCLL

Specifications

Accession Number AAH21101.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9440
Molecular Weight (g/mol) 99.3kDa
Name MED17 (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MSGVRAVRISIESACEKQVHEVGLDGTETYLPPLSMSQNLARLAQRIDFSQGSGSEEEEAAGTEGDAQDWPGAGSSADQDDEEGVVKFQPSLWPWDSVRNNLRSALTEMCVLYDVLSIVRDKKFMTLDPVSQDALPPKQNPQTLQLISKKKSLAGAAQILLKGAERLTKSVTENQENKLQRDFNSELLRLRQHWKLRKVGDKILGDLSYRSAGSLFPHHGTFEVIKNTDLDLDKKIPEDYCPLDVQIPSDLEGSAYIKVSIQKQAPDIGDLGTVNLFKRPLPKSKPGSPHWQTKLEAAQNVLLCKEIFAQLSREAVQIKSQVPHIVVKNQIISQPFPSLQLSISLCHSSNDKKSQKFATEKQCPEDHLYVLEHNLHLLIREFHKQTLSSIMMPHPASAPFGHKRMRLSGPQAFDKNEINSLQSSEGLLEKIIKQAKHIFLRSRAAATIDSLASRIEDPQIQAHWSNINDVYESSVKVLITSQGYEQICKSIQLQLNIGVEQIRVVHRDGRVITLSYQEQELQDFLLSQMSQHQVHAVQQLAKVMGWQVLSFSNHVGLGPIESIGNASAITVASPSGDYAISVRNGPESGSKIMVQFPRNQCKDLPKSDVLQDNKWSHLRGPFKEVQWNKMEGRNFVYKMELLMSALSPCLL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CRSP6/CRSP77/DRIP80/FLJ10812/TRAP80
Common Name MED17
Gene Symbol MED17
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.