Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MED4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5089
This item is not returnable.
View return policy
Description
MED4 Polyclonal specifically detects MED4 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MED4 | |
Polyclonal | |
Unconjugated | |
Q9NPJ6 | |
MED4 | |
E. coli-derived recombinant human MED4 Met80-Asp261 Accession # Q9NPJ6 | |
100 μg | |
Primary | |
Detects endogenous human MED4 in Western blots. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Activator-recruited cofactor 36 kDa component, ARC36, DRIP36, DRIP36mediator of RNA polymerase II transcription, subunit 4 homolog (S. cerevisiae), FLJ10956, HSPC126, HSPC126vitamin D receptor interacting protein, mediator complex subunit 4TRAP/SMCC/PC2 subunit p36 subunit, subunit 4 homolog, TRAP36, VDRIP | |
Goat | |
Affinity Purified | |
RUO | |
29079 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction