Learn More
Human Megalin/LRP2 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | LRP2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 545606 |
| Conjugate | Alexa Fluor 594 |
| Dilution | Flow Cytometry 0.25-1μg/106 cells |
| Formulation | Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | P98164 |
| Gene Alias | calcium sensor protein, DBS, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP2, LRP-2, megalin |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.