Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Megalin/LRP2 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB9578S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9578S-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB9578S-100 Supplier R&D Systems Supplier No. FAB9578S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LRP2 Monoclonal specifically detects LRP2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen LRP2
Applications Flow Cytometry
Classification Monoclonal
Clone 545606
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1μg/106 cells
Formulation Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P98164
Gene Alias calcium sensor protein, DBS, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP2, LRP-2, megalin
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Megalin/LRP2 Pro3510-Lys3964 Accession # P98164
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4036
Test Specificity Detects human Megalin/LRP2 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.