Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Melan-A/MART-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8008
This item is not returnable.
View return policy
Description
Melan-A/MART-1 Monoclonal specifically detects Melan-A/MART-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Melan-A/MART-1 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q16655 | |
MLANA | |
E. coli-derived recombinant human Melan-A/MART-1 Asn52-Pro118 Accession # Q16655 | |
100 μg | |
Primary | |
Detects human Melan-A/MART-1 in ELISA. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
872719 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Antigen LB39-AA, Antigen SK29-AA, Mart 1 Melan A, MART-1, MART1MART-1, melan-A, MelanA, melanoma antigen recognized by T-cells 1, MLANA, Protein Melan-A | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2315 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction