Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Melan-A/MART-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF8008
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8008 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF8008 Supplier R&D Systems Supplier No. AF8008
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Melan-A/MART-1 Polyclonal specifically detects Melan-A/MART-1 in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Melan-A/MART-1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL
Gene Accession No. Q16655
Gene Alias Antigen LB39-AA, Antigen SK29-AA, Mart 1 Melan A, MART-1, MART1MART-1, melan-A, MelanA, melanoma antigen recognized by T-cells 1, MLANA, Protein Melan-A
Gene Symbols MLANA
Host Species Sheep
Immunogen E. coli-derived recombinant human Melan-A/MART-1 Asn52-Pro118 Accession # Q16655
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2315
Test Specificity Detects human Melan-A/MART-1 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.