Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Melanopsin Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB10497U100UG
This item is not returnable.
View return policy
Description
Melanopsin Monoclonal specifically detects Melanopsin in Human samples. It is validated for Flow Cytometry.Specifications
Melanopsin | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
melanopsin, MGC142118, MOP, OPN4, opsin 4, Opsin-4 | |
NS0 mouse myeloma cell line transfected with human Melanopsin, Accession # Q9UHM6 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
628737 | |
Flow Cytometry 0.25-1 ug/106 cells | |
Q9UHM6 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
94233 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction