Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Melanotransferrin/CD228 Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA81751U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA81751U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA81751U100 Supplier R&D Systems Supplier No. FAB81751U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Melanotransferrin/CD228/MFI2 Monoclonal specifically detects Melanotransferrin/CD228/MFI2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Melanotransferrin/CD228/MFI2
Applications Flow Cytometry
Classification Monoclonal
Clone 893416
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P08582
Gene Alias antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and96.5, CD228, CD228 antigen, FLJ38863, MAP97, MAP97Melanoma-associated antigen p97, melanotransferrin, membrane-bound transferrin-like protein, MFI2, MGC4856, MTF, MTF1, p97
Gene Symbols MFI2
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Melanotransferrin/CD228 Gly20-Gly711 Accession # P08582
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4241
Test Specificity Detects human Melanotransferrin/CD228 in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.