Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Melanotransferrin/CD228 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8175SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8175SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8175SP Supplier R&D Systems Supplier No. MAB8175SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Melanotransferrin/CD228/MFI2 Monoclonal specifically detects Melanotransferrin/CD228/MFI2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen Melanotransferrin/CD228/MFI2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 893438
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P08582
Gene Alias antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and96.5, CD228, CD228 antigen, FLJ38863, MAP97, MAP97Melanoma-associated antigen p97, melanotransferrin, membrane-bound transferrin-like protein, MFI2, MGC4856, MTF, MTF1, p97
Gene Symbols MFI2
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Melanotransferrin/CD228 Met1-Gly711 Accession # P08582
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4241
Test Specificity Detects human Melanotransferrin/CD228 in ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.