Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MEPE/OF45 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3140
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3140 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3140 Supplier R&D Systems Supplier No. AF3140
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

MEPE/OF45 Polyclonal specifically detects MEPE/OF45 in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen MEPE/OF45
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 2-6 ug/mL
Gene Accession No. Q9NQ76
Gene Alias extracellular phosphoglycoprotein with ASARM motif (bone), matrix extracellular phosphoglycoprotein, OF45, Osteoregulin
Gene Symbols MEPE
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human MEPE/OF45 Pro18-Asp525 Accession # Q9NQ76
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56955
Test Specificity Detects human MEPE/OF45 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.