Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Meprin alpha Subunit/MEP1A Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC3220U100UG
This item is not returnable.
View return policy
Description
Meprin alpha Subunit/MEP1A Monoclonal specifically detects Meprin alpha Subunit/MEP1A in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Meprin alpha Subunit/MEP1A | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
AAA21338 | |
MEP1A | |
Mouse myeloma cell line NS0-derived recombinant human Meprin alpha Subunit/MEP1A Val22-Gln601 Accession # AAA21338 | |
RUO | |
4224 | |
Human | |
IgG2b |
Flow Cytometry | |
364312 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
bA268F1.1 (meprin A alpha (PABA peptide hydrolase)), EC 3.4.24, EC 3.4.24.18, endopeptidase-2, MEP1A, meprin A subunit alpha, meprin A, alpha (PABA peptide hydrolase), N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha, PABA peptide hydrolase, PPH alpha, PPHA | |
Mouse | |
100 μg | |
Primary | |
Detects human Meprin alpha Subunit/MEP1A in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human MEP1B or recombinant mouse MEP1A is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction