Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Mer Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
$254.50 - $2240.00
Specifications
Antigen | Mer |
---|---|
Clone | 125518 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Flow Cytometry, CyTOF, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8912SP
![]() |
R&D Systems
MAB8912SP |
25 μg |
Each for $254.50
|
|
|||||
MAB8912100
![]() |
R&D Systems
MAB8912100 |
100 μg |
Each for $631.50
|
|
|||||
MAB8912500
![]() |
R&D Systems
MAB8912500 |
500 μg |
Each for $2,240.00
|
|
|||||
Description
Mer Monoclonal specifically detects Mer in Human samples. It is validated for Flow Cytometry, CyTOF-ready, Knockout Validated, ELISA Capture (Matched Antibody Pair).Specifications
Mer | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
AAB60430 | |
10461 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Mer Met1-Ala499 Accession # AAB60430 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Mer in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Axl, rhDtk, or recombinant mouse Mer is observed. |
125518 | |
Flow Cytometry, CyTOF, ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
c-Eyk, C-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MerTK, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer | |
MERTK | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title