Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Mer Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$214.00 - $702.00
Specifications
Antigen | Mer |
---|---|
Clone | 125508 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Mer Monoclonal specifically detects Mer in Human samples. It is validated for Western Blot.Specifications
Mer | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
c-Eyk, C-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MerTK, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer | |
MERTK | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
125508 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q12866.2 | |
10461 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Mer Ala21-Ala499 Accession # Q12866.2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Mer in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) Axl, rhDtk, or rmMer. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title