Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Mesothelin Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB32652G100UG
This item is not returnable.
View return policy
Description
Mesothelin Monoclonal specifically detects Mesothelin in Human samples. It is validated for Flow Cytometry.Specifications
Mesothelin | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAH09272 | |
MSLN | |
Mouse myeloma cell line NS0-derived recombinant human Mesothelin Glu296-Gly580 Accession # AAH09272 | |
RUO | |
10232 | |
Human | |
IgG2a |
Flow Cytometry | |
420411 | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPF, MPFSMRP, MSLN, Pre-pro-megakaryocyte-potentiating factor, SMR, soluble MPF mesothelin related protein | |
Rat | |
100 μg | |
Primary | |
Detects human Mesothelin in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction