Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Mesothelin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$173.10 - $464.00
Specifications
Antigen | Mesothelin |
---|---|
Clone | 618924 |
Dilution | Immunocytochemistry 8-25 μg/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11417100
![]() |
R&D Systems
MAB11417100 |
100 μg |
Each for $464.00
|
|
|||||
MAB11417SP
![]() |
R&D Systems
MAB11417SP |
25 μg |
Each for $173.10
|
|
|||||
Description
Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for ImmunocytochemistrySpecifications
Mesothelin | |
Immunocytochemistry 8-25 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPF, MPFSMRP, MSLN, Pre-pro-megakaryocyte-potentiating factor, SMR, soluble MPF mesothelin related protein | |
Mouse myeloma cell line, NS0-derived human Mesothelin, Glu296-Gly580, Accession # Q13421 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
618924 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q13421 | |
10232 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title