Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Mesothelin PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 6 publications
Supplier: R&D Systems FAB32652P
This item is not returnable.
View return policy
Description
Mesothelin Monoclonal specifically detects Mesothelin in Human samples. It is validated for Flow Cytometry, Knockout Validated.Specifications
Mesothelin | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPF, MPFSMRP, MSLN, Pre-pro-megakaryocyte-potentiating factor, SMR, soluble MPF mesothelin related protein | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10232 | |
Human | |
Purified |
Flow Cytometry, Immunoassay | |
420411 | |
Flow Cytometry 10 uL/10^6 cells, Knockout Validated 10 uL/10^6 cells | |
AAH09272 | |
MSLN | |
Mouse myeloma cell line NS0-derived recombinant human Mesothelin Glu296-Gly580 Accession # AAH09272 | |
100 Tests | |
Primary | |
Detects human Mesothelin in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction