Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MESP1 Alexa Fluor 750-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Supplier: R&D Systems IC9219S100UG
This item is not returnable.
View return policy
Description
Mesp1 Monoclonal specifically detects Mesp1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| Mesp1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide. | |
| BHLHC5, bHLHc5MGC10676, Class C basic helix-loop-helix protein 5, mesoderm posterior 1 homolog (mouse), mesoderm posterior protein 1 | |
| E. coli-dervived recombinant human MESP1, Met1-Gln85, Accession No. Q9BRJ9 | |
| 100 μg | |
| 55897.0 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 2030b | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9BRJ9 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human MESP1 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction