Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MESP1 Antibody

Catalog No. MAB92192100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB92192100 100 μg
MAB92192SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB92192100 Supplier R&D Systems Supplier No. MAB92192100
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mesp1 Monoclonal specifically detects Mesp1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Mesp1
Applications Western Blot
Classification Monoclonal
Clone 939816
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q9BRJ9
Gene Alias BHLHC5, bHLHc5MGC10676, Class C basic helix-loop-helix protein 5, mesoderm posterior 1 homolog (mouse), mesoderm posterior protein 1
Host Species Mouse
Immunogen E. coli-derived recombinant human MESP1, Met1-Gln85, Accession No. Q9BRJ9
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 55897.0
Test Specificity Detects human MESP1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.