Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MET (P08581, 18 a.a. - 217 a.a.) Partial Recombinant Protein

Catalog No. 89952198
Click to view available options
Quantity:
10 μg

Used for SDS-PAGE

The proto-oncogene MET product is the hepatocyte growth factor receptor and encodes tyrosine-kinase activity. The primary single chain precursor protein is post-translationally cleaved to produce the alpha and beta subunits, which are disulfide linked to form the mature receptor. Various mutations in the MET gene are associated with papillary renal carcinoma. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Accession Number P08581
Concentration 0.1mg/mL
For Use With (Application) SDS-PAGE
Formulation In 50mM Tris-HCl, 10mM GSH, pH 7.5
Gene ID (Entrez) 4233
Molecular Weight (g/mol) 50kDa
Name Human MET (P08581, 18 a.a. - 217 a.a.) Partial Recombinant Protein
Quantity 10 μg
Storage Requirements Store at −80°C.Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias AUTS9, HGFR, RCCP2, c-Met
Common Name MET
Gene Symbol MET
Species E. coli
Recombinant Recombinant
Protein Tag Untagged
Expression System Escherichia coli expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.