Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Methionine Aminopeptidase 1/METAP1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3537
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3537 100 μg
MAB3537SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3537 Supplier R&D Systems Supplier No. MAB3537
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Methionine Aminopeptidase 1/METAP1 Monoclonal specifically detects Methionine Aminopeptidase 1/METAP1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Methionine Aminopeptidase 1/METAP1
Applications Western Blot
Classification Monoclonal
Clone 405419
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P53582
Gene Alias DKFZp781C0419, EC 3.4.11.18, KIAA0094MAP 1, MAP1, MAP1A, MetAP 1, MetAP1, MetAP1A, methionine aminopeptidase 1, methionyl aminopeptidase 1, Peptidase M 1
Gene Symbols METAP1
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Methionine Aminopeptidase 1/METAP1 His52-Phe386 Accession # P53582
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23173
Test Specificity Detects human Methionine Aminopeptidase 1/METAP1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Methionine Aminopeptidase 2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.