Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Methionine Aminopeptidase 1D/MAP1D Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4714
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4714 100 μg
AF4714SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF4714 Supplier R&D Systems Supplier No. AF4714
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Methionine Aminopeptidase 1D/MAP1D Polyclonal specifically detects Methionine Aminopeptidase 1D/MAP1D in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Methionine Aminopeptidase 1D/MAP1D
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. Q6UB28
Gene Alias EC 3.4.11.18, MAP1D, MAP1DCDS of metAP-3 within PCR fragment, MetAP1D, Metap1l, methionine aminopeptidase 1D, mitochondrial, methionyl aminopeptidase type 1D (mitochondrial), Methionyl aminopeptidase type 1D, mitochondrial
Gene Symbols METAP1D
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Methionine Aminopeptidase 1D/MAP1D Arg44-Ala335 Accession # Q6UB28
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254042
Test Specificity Detects human Methionine Aminopeptidase 1D/MAP1D in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.