Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MFG-E8 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC27671G100UG
This item is not returnable.
View return policy
Description
MFG-E8 Monoclonal specifically detects MFG-E8 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
MFG-E8 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q08431 | |
MFGE8 | |
Mouse myeloma cell line NS0-derived recombinant human MFG-E8 Leu24-Cys387 Accession # Q08431 | |
100 μg | |
Primary | |
Detects human MFG-E8 in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Flow Cytometry | |
278918 | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
BA46, Breast epithelial antigen BA46, EDIL1, hP47, HsT19888, lactadherin, Lactahedrin, Medin, MFG1, MFGE8, MFG-E8, MFGM, Milk fat globule-EGF factor 8, milk fat globule-EGF factor 8 protein, O-acetyl disialoganglioside synthase, OAcGD3S, SED1, SPAG10, sperm associated antigen 10, sperm surface protein hP47 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4240 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction