Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MFG-E8 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF2767
This item is not returnable.
View return policy
Description
MFG-E8 Polyclonal specifically detects MFG-E8 in Human samples. It is validated for Western Blot, Simple Western.Specifications
MFG-E8 | |
Polyclonal | |
Unconjugated | |
Q08431 | |
MFGE8 | |
Mouse myeloma cell line NS0-derived human MFG-E8 Leu24-Cys387 Accession # Q08431 | |
100 μg | |
Primary | |
Detects human MFG-E8 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
BA46, Breast epithelial antigen BA46, EDIL1, hP47, HsT19888, lactadherin, Lactahedrin, Medin, MFG1, MFGE8, MFG-E8, MFGM, Milk fat globule-EGF factor 8, milk fat globule-EGF factor 8 protein, O-acetyl disialoganglioside synthase, OAcGD3S, SED1, SPAG10, sperm associated antigen 10, sperm surface protein hP47 | |
Sheep | |
Affinity Purified | |
RUO | |
4240 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction