Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MFRP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1915
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1915 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1915 Supplier R&D Systems Supplier No. MAB1915
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

MFRP Monoclonal specifically detects MFRP in Human samples. It is validated for Western Blot.

Specifications

Antigen MFRP
Applications Western Blot
Classification Monoclonal
Clone 291218
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9BY79
Gene Alias MCOP5, membrane frizzled-related protein, Membrane-type frizzled-related protein, MGC32938, NNO2, RD6
Gene Symbols MFRP
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human MFRP Ser101-Pro579 Accession # Q9BY79
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83552
Test Specificity Detects human MFRP in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) Frizzled-1, -2, -3, -4, -6, -7, -8, -9, recombinant human Frizzled-5, or rmMFRP is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.