Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human mGluR1 Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB48362100
This item is not returnable.
View return policy
Description
mGluR1 Monoclonal antibody specifically detects mGluR1 in Human samples. It is validated for ImmunohistochemistrySpecifications
mGluR1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
glutamate receptor, metabotropic 1, GPRC1A, GRM1, GRM1A, metabotropic glutamate receptor 1, mGluR1, MGLUR1A, MGLUR1GPRC1AmGlu1 | |
Synthetic peptide, Accession # P42261 | |
100 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG2b |
Immunohistochemistry | |
1059503 | |
Immunohistochemistry 5-25 ug/mL | |
P42261 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2911 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction