Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human mGluR2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $444.50
Specifications
Antigen | mGluR2 |
---|---|
Clone | 455311 |
Dilution | Immunohistochemistry 5-25 μ/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB46761SP
![]() |
R&D Systems
MAB46761SP |
25 μg |
Each for $200.50
|
|
|||||
MAB46761100
![]() |
R&D Systems
MAB46761100 |
100 μg |
Each for $444.50
|
|
|||||
Description
mGluR2 Monoclonal specifically detects mGluR2 in Human samples. It is validated for Immunohistochemistry.Specifications
mGluR2 | |
Immunohistochemistry 5-25 μ/mL | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
GLUR2, glutamate metabotropic receptor 2, glutamate receptor, metabotropic 2, GPRC1B, GPRC1Bmetabotropic glutamate receptor 2, GRM2, mGlu2, mGluR2, MGLUR2glutamate receptor homolog, mGluR7 | |
Mouse myeloma cell line, NS0-derived human mGluR2, Glu19-Ser498, Accession No. Q14416 | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
455311 | |
Immunohistochemistry | |
Unconjugated | |
Human | |
Q14416 | |
2912 | |
IgG2b | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human mGluR2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title