Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human mGluR5 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC4514V100UG
This item is not returnable.
View return policy
Description
mGluR5 Monoclonal specifically detects mGluR5 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
mGluR5 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P41594 | |
GRM5 | |
Mouse myeloma cell line NS0-derived recombinant human mGluR5 Gln21-Ser509 Accession # P41594 | |
RUO | |
2915 | |
Human | |
IgG1 |
Flow Cytometry | |
464818 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
glutamate receptor, metabotropic 5, GPRC1E, GPRC1EMGLUR5metabotropic glutamate receptor 5, GRM5, mGlu5, mGluR5 | |
Mouse | |
100 μg | |
Primary | |
Detects human mGluR5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human mGluR1, R2, R3, R4, R7, or R8 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction